Assay Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009209.3 | NP_033235.3 | 8 - 9 | 1464 | 89 | |
| GenBank mRNA | AJ439987.1 | - | 8 - 9 | 1281 | 89 | |
| AY188506.1 | - | 8 - 9 | 1281 | 89 | ||
| AY525777.1 | - | 8 - 9 | 1578 | 89 | ||
| AY525778.1 | - | 8 - 9 | 1578 | 89 | ||
| BC066216.1 | - | 9 - 10 | 1398 | 30431573 | 89 | |
| U76306.1 | - | 8 - 9 | 1263 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a2
Genomic Map