Assay Details
| Gene Symbol: Slc6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009209.3 | NP_033235.3 | 10 - 11 | 1689 | 77 | |
| GenBank mRNA | AJ439987.1 | - | 10 - 11 | 1506 | 77 | |
| AY188506.1 | - | 10 - 11 | 1506 | 77 | ||
| AY525777.1 | - | 10 - 11 | 1803 | 77 | ||
| AY525778.1 | - | 10 - 11 | 1803 | 77 | ||
| U76306.1 | - | 10 - 11 | 1488 | 77 | ||
More Information
Set Membership: |
|

Slc6a2
Genomic Map