Assay Details
| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009255.4 | NP_033281.1 | 1 - 2 | 156 | 89 | |
| GenBank mRNA | AK045954.1 | - | 1 - 2 | 128 | 89 | |
| AK047856.1 | - | 1 - 2 | 128 | 89 | ||
| AK051332.1 | - | 1 - 2 | 128 | 89 | ||
| AK159687.1 | - | 1 - 2 | 128 | 89 | ||
| AK160700.1 | - | 1 - 2 | 128 | 89 | ||
| AK164270.1 | - | 1 - 2 | 128 | 89 | ||
| BC010675.1 | - | 1 - 2 | 106 | 3595407 | 89 | |
| X70296.1 | - | 1 - 2 | 127 | 89 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK045954.1,AK047856.1,AK051332.1,AK159687.1,BC010675.1,NM_009255.4,X70296.1 transcripts.
Set Membership: |
|

Serpine2
Genomic Map