Assay Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009271.3 | NP_033297.2 | 5 - 6 | 729 | 59 | |
| GenBank mRNA | M17031.1 | - | 2 - 3 | 345 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Src
Genomic Map