Assay Details
| Gene Symbol: Src | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009271.3 | NP_033297.2 | 6 - 7 | 747 | 75 | |
| GenBank mRNA | M17031.1 | - | 3 - 4 | 363 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Src
Genomic Map