Assay Details
| Gene Symbol: Srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009275.4 | NP_033301.1 | 3 - 4 | 325 | 121 | |
| GenBank mRNA | AK004854.1 | - | 3 - 4 | 358 | 121 | |
| AK007859.1 | - | 3 - 4 | 331 | 121 | ||
| AK008383.1 | - | 3 - 4 | 368 | 121 | ||
| AK015469.1 | - | 3 - 4 | 390 | 121 | ||
| AK030437.1 | - | 3 - 4 | 326 | 121 | ||
| AK075583.1 | - | 3 - 4 | 328 | 121 | ||
| AK167039.1 | - | 3 - 4 | 331 | 121 | ||
| BC003798.1 | - | 3 - 4 | 346 | 3490766 | 121 | |
| U17343.1 | - | 3 - 4 | 339 | 121 | ||
More Information
Set Membership: |
|

Srprb
Genomic Map