Assay Details
| Gene Symbol: Stxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113569.1 | NP_001107041.1 | 11 - 12 | 1110 | 115 | |
| NM_009295.2 | NP_033321.2 | 11 - 12 | 1105 | 115 | ||
| GenBank mRNA | AF326545.1 | - | 11 - 12 | 964 | 115 | |
| AF326563.1 | - | 11 - 12 | 964 | 115 | ||
| AK004780.1 | - | 6 - 7 | 829 | 115 | ||
| BC031728.1 | - | 11 - 12 | 1037 | 4480978 | 115 | |
| D45903.1 | - | 11 - 12 | 999 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stxbp1
Genomic Map