Assay Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252341.1 | NP_001239270.1 | 1 - 2 | 399 | 89 | |
| GenBank mRNA | AK078790.1 | - | 1 - 2 | 371 | 89 | |
| D37792.1 | - | 1 - 2 | 263 | 89 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK078790.1,D37792.1,NM_001252341.1 transcripts.
Set Membership: |
|

Syt1
Genomic Map