Assay Details
| Gene Symbol: Syt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252341.1 | NP_001239270.1 | 8 - 9 | 1308 | 73 | |
| NM_001252342.1 | NP_001239271.1 | 7 - 8 | 1219 | 73 | ||
| NM_009306.3 | NP_033332.1 | 7 - 8 | 1228 | 73 | ||
| GenBank mRNA | AK018163.1 | - | 7 - 8 | 1144 | 73 | |
| AK078790.1 | - | 8 - 9 | 1280 | 73 | ||
| AK164153.1 | - | 7 - 8 | 1120 | 73 | ||
| BC042519.1 | - | 7 - 8 | 872 | 5365461 | 73 | |
| BC048187.1 | - | 7 - 8 | 1206 | 73 | ||
| D37792.1 | - | 8 - 9 | 1168 | 73 | ||
| FR827897.1 | - | 5 - 6 | 709 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt1
Genomic Map