Assay Details
| Gene Symbol: Syt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009307.3 | NP_033333.2 | 7 - 8 | 1196 | 62 | |
| GenBank mRNA | AF257304.1 | - | 6 - 7 | 951 | 62 | |
| AK036357.1 | - | 7 - 8 | 1197 | 62 | ||
| AK220537.1 | - | 4 - 5 | 510 | 62 | ||
| BC027019.1 | - | 6 - 7 | 957 | 5362352 | 62 | |
| D37793.1 | - | 5 - 6 | 830 | 62 | ||
More Information
Set Membership: |
|

Syt2
Genomic Map