Assay Details
| Gene Symbol: Tac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009311.2 | NP_033337.1 | 2 - 3 | 227 | 106 | |
| GenBank mRNA | BC117081.1 | - | 2 - 3 | 202 | 40126400 | 106 |
| BC119122.1 | - | 2 - 3 | 202 | 8733871 | 106 | |
| BC144737.1 | - | 2 - 3 | 202 | 9053285 | 106 | |
| D17584.1 | - | 2 - 3 | 227 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tac1
Genomic Map