Assay Details
| Gene Symbol: Tac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199971.1 | NP_001186900.1 | 1 - 2 | 202 | 88 | |
| GenBank mRNA | D14423.1 | - | 1 - 2 | 202 | 88 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the D14423.1,NM_001199971.1 transcripts.
Set Membership: |
|

Tac2
Genomic Map