Assay Details
| Gene Symbol: Tac2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199971.1 | NP_001186900.1 | 4 - 5 | 475 | 78 | |
| NM_009312.2 | NP_033338.2 | 3 - 4 | 303 | 78 | ||
| GenBank mRNA | AF186116.1 | - | 3 - 4 | 269 | 78 | |
| BC031348.1 | - | 3 - 4 | 300 | 78 | ||
| BC051476.1 | - | 3 - 4 | 309 | 1533442 | 78 | |
| BQ175657.1 | - | 4 - 5 | 78 | |||
| D14423.1 | - | 4 - 5 | 475 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tac2
Genomic Map