Assay Details
| Gene Symbol: Tacr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009313.5 | NP_033339.2 | 4 - 5 | 1649 | 67 | |
| GenBank mRNA | AK038558.1 | - | 3 - 4 | 1306 | 67 | |
| BC075631.1 | - | 4 - 5 | 1649 | 30536972 | 67 | |
| BC098332.1 | - | 3 - 4 | 1556 | 30618488 | 67 | |
| L27826.1 | - | 3 - 4 | 547 | 67 | ||
| L27828.1 | - | 3 - 4 | 547 | 67 | ||
| X62934.1 | - | 3 - 4 | 753 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tacr1
Genomic Map