Assay Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009373.3 | NP_033399.1 | 2 - 3 | 278 | 83 | |
| GenBank mRNA | AF076928.1 | - | 1 - 2 | 193 | 83 | |
| AK052912.1 | - | 2 - 3 | 253 | 83 | ||
| AK054108.1 | - | 2 - 3 | 253 | 83 | ||
| AK080224.1 | - | 2 - 3 | 278 | 83 | ||
| AK080593.1 | - | 2 - 3 | 253 | 83 | ||
| AK086001.1 | - | 2 - 3 | 253 | 83 | ||
| AK089481.1 | - | 2 - 3 | 281 | 83 | ||
| AK143712.1 | - | 2 - 3 | 314 | 83 | ||
| AK151776.1 | - | 2 - 3 | 279 | 83 | ||
| AK152152.1 | - | 2 - 3 | 253 | 83 | ||
| AK152627.1 | - | 2 - 3 | 279 | 83 | ||
| AK159255.1 | - | 2 - 3 | 253 | 83 | ||
| AK166302.1 | - | 2 - 3 | 253 | 83 | ||
| AK168990.1 | - | 2 - 3 | 254 | 83 | ||
| AK169356.1 | - | 2 - 3 | 281 | 83 | ||
| BC016492.1 | - | 2 - 3 | 219 | 3256943 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tgm2
Genomic Map