Assay Details
| Gene Symbol: Tial1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009383.2 | NP_033409.1 | 7 - 8 | 967 | 110 | |
| GenBank mRNA | AK012085.1 | - | 7 - 8 | 611 | 110 | |
| BC010496.1 | - | 8 - 9 | 701 | 3497971 | 110 | |
| BC068194.1 | - | 3 - 4 | 243 | 110 | ||
| U55861.1 | - | 7 - 8 | 743 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tial1
Genomic Map