Assay Details
| Gene Symbol: Tial1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009383.2 | NP_033409.1 | 10 - 11 | 1268 | 87 | |
| GenBank mRNA | AK012085.1 | - | 10 - 11 | 912 | 87 | |
| AK132677.1 | - | 11 - 12 | 1436 | 87 | ||
| BC010496.1 | - | 11 - 12 | 1002 | 3497971 | 87 | |
| BC068194.1 | - | 6 - 7 | 544 | 87 | ||
| U55861.1 | - | 10 - 11 | 1044 | 87 | ||
More Information
Set Membership: |
|

Tial1
Genomic Map