Assay Details
| Gene Symbol: Tnnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009393.2 | NP_033419.1 | 1 - 2 | 66 | 89 | |
| GenBank mRNA | BC061172.1 | - | 1 - 2 | 60 | 30300744 | 89 |
| M29793.1 | - | 1 - 2 | 66 | 89 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC061172.1,M29793.1,NM_009393.2 transcripts.
Set Membership: |
|

Tnnc1
Genomic Map