Assay Details
| Gene Symbol: Tnfrsf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009400.2 | NP_033426.1 | 1 - 2 | 194 | 89 | |
| NM_021985.2 | NP_068820.1 | 1 - 2 | 194 | 89 | ||
| GenBank mRNA | AF229432.1 | - | 1 - 2 | 194 | 89 | |
| AF229433.1 | - | 1 - 2 | 194 | 89 | ||
| AF229434.1 | - | 1 - 2 | 194 | 89 | ||
| AK020762.1 | - | 1 - 2 | 215 | 89 | ||
| AK081878.1 | - | 1 - 2 | 183 | 89 | ||
| AY157832.1 | - | 1 - 2 | 194 | 89 | ||
| AY157833.1 | - | 1 - 2 | 194 | 89 | ||
| U82534.1 | - | 1 - 2 | 194 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441978 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025490 | U82534.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025490 | AF229433.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025490 | AF229434.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025490 | NM_009400.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2025490 | NM_021985.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 294689 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166814 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110329 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025490 | U82534.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025490 | AF229433.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025490 | AF229434.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025490 | NM_009400.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2025490 | NM_021985.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469401 | U82534.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469401 | AF229433.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469401 | AF229434.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469401 | AK020762.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P469401 | NM_009400.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441922 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441902 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441920 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441907 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441912 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441905 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441903 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441894 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441931 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110329 | - | - |
| Whole Mouse Genome G4122A | A_51_P469401 | U82534.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469401 | AF229433.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469401 | AF229434.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469401 | AK020762.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P469401 | NM_009400.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120726 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110329 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04089299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04089283 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04089289 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04089307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04089282 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441979 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441978 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441977 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23441976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441893 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441897 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441916 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03398886 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03854211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03768328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03603827 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441912 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04089298 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441931 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441900 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03854214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03398887 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04089301 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03202353 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441894 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03961264 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04089304 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03992745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04089303 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23441925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10511296 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511294 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511293 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511292 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511297 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511295 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10511291 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422303_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10511290 | - | - |
| Mouse Genome 430 2.0 Array | 1422303_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1422303_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21936_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21936_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400017738.mm.1 | NM_009400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017738.mm.1 | NM_009400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017738.mm.1 | NM_021985.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017738.mm.1 | NM_021985.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017738.mm.1 | TC0400002104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400017740.mm.1 | NM_009400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017740.mm.1 | NM_009400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017740.mm.1 | NM_021985.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017740.mm.1 | NM_021985.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017740.mm.1 | TC0400002104.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400017736.mm.1 | NM_009400.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017736.mm.1 | NM_009400.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017736.mm.1 | NM_021985.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017736.mm.1 | NM_021985.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400017736.mm.1 | TC0400002104.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1422303_PM_a_at | - | - |
| Mouse Expression Set 430 | 1422303_a_at | - | - |
| Murine Genome U74v2 Set | 104662_at | - | - |
| Murine Genome U74v2 Set | 104662_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422303_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422303_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422303_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422303_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 316156 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3170746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3170746 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 430253 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003170746 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000430253 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2465751 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2419490 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2419490 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2465751 | - | - |
| mouse-6 v1.1 (Illumina) | scl24601.2.1_6-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0002685.1_109-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3170746 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 430253 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002685.1_109-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24601.2.1_6-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430253 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3170746 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2419490 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2419494 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2419494 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2419490 | - | - |

Tnfrsf18
Genomic Map