Assay Details
| Gene Symbol: Tnni3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009406.3 | NP_033432.1 | 4 - 5 | 281 | 61 | |
| GenBank mRNA | AK169223.1 | - | 4 - 5 | 282 | 61 | |
| BC061171.1 | - | 4 - 5 | 281 | 30300830 | 61 | |
| BC100590.1 | - | 4 - 5 | 212 | 6787916 | 61 | |
| U09181.1 | - | 2 - 3 | 153 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnni3
Genomic Map