Assay Details
| Gene Symbol: Ttk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110265.1 | NP_001103735.1 | 11 - 12 | 1317 | 82 | |
| NM_001284272.1 | NP_001271201.1 | 11 - 12 | 1317 | 82 | ||
| NM_009445.2 | NP_033471.2 | 11 - 12 | 1317 | 82 | ||
| GenBank mRNA | AK041487.1 | - | 11 - 12 | 1288 | 82 | |
| AK049200.1 | - | 11 - 12 | 1312 | 82 | ||
| AK164093.1 | - | 11 - 12 | 1316 | 82 | ||
| AX775731.1 | - | 11 - 12 | 1283 | 82 | ||
| BC058851.1 | - | 11 - 12 | 1303 | 30023533 | 82 | |
| M86377.1 | - | 12 - 13 | 1343 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttk
Genomic Map