| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500026637.mm.1 |
TC0500003000.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026637.mm.1 |
NM_009469.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026637.mm.1 |
NM_009469.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026636.mm.1 |
TC0500003000.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026636.mm.1 |
NM_009469.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026636.mm.1 |
NM_009469.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026635.mm.1 |
TC0500003000.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500026635.mm.1 |
NM_009469.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500026635.mm.1 |
NM_009469.3 |
Refseq |