Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009469.3 | NP_033495.2 | 13 - 14 | 1451 | 117 | |
| GenBank mRNA | AF053756.1 | - | 13 - 14 | 1143 | 117 | |
| AF072370.1 | - | 13 - 14 | 1134 | 117 | ||
| AK122357.1 | - | 12 - 13 | 953 | 117 | ||
| AK148317.1 | - | 13 - 14 | 1426 | 117 | ||
| AK154939.1 | - | 13 - 14 | 1468 | 117 | ||
| AK156534.1 | - | 13 - 14 | 1428 | 117 | ||
| BC053062.1 | - | 13 - 14 | 1111 | 117 | ||
| BC057121.1 | - | 13 - 14 | 1451 | 6834534 | 117 | |
| BC059835.1 | - | 13 - 14 | 1402 | 6406755 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ulk1
Genomic Map