Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009469.3 | NP_033495.2 | 17 - 18 | 1876 | 91 | |
| GenBank mRNA | AF053756.1 | - | 17 - 18 | 1568 | 91 | |
| AF072370.1 | - | 17 - 18 | 1559 | 91 | ||
| AK122357.1 | - | 16 - 17 | 1378 | 91 | ||
| AK154939.1 | - | 17 - 18 | 1893 | 91 | ||
| AK156534.1 | - | 17 - 18 | 1853 | 91 | ||
| BC053062.1 | - | 17 - 18 | 1536 | 91 | ||
| BC057121.1 | - | 17 - 18 | 1876 | 6834534 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ulk1
Genomic Map