Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009469.3 | NP_033495.2 | 18 - 19 | 1962 | 86 | |
| GenBank mRNA | AF053756.1 | - | 18 - 19 | 1654 | 86 | |
| AF072370.1 | - | 18 - 19 | 1645 | 86 | ||
| AK122357.1 | - | 17 - 18 | 1464 | 86 | ||
| AK148317.1 | - | 18 - 19 | 1955 | 86 | ||
| AK154939.1 | - | 18 - 19 | 1979 | 86 | ||
| BC053062.1 | - | 18 - 19 | 1622 | 86 | ||
| BC057121.1 | - | 18 - 19 | 1962 | 6834534 | 86 | |
| BC059835.1 | - | 18 - 19 | 1931 | 6406755 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ulk1
Genomic Map