Assay Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009469.3 | NP_033495.2 | 19 - 20 | 2234 | 123 | |
| GenBank mRNA | AF053756.1 | - | 19 - 20 | 1926 | 123 | |
| AF072370.1 | - | 19 - 20 | 1917 | 123 | ||
| AK122357.1 | - | 18 - 19 | 1736 | 123 | ||
| AK148317.1 | - | 19 - 20 | 2227 | 123 | ||
| AK154939.1 | - | 19 - 20 | 2251 | 123 | ||
| AK156534.1 | - | 19 - 20 | 2211 | 123 | ||
| BC053062.1 | - | 19 - 20 | 1894 | 123 | ||
| BC057121.1 | - | 19 - 20 | 2234 | 6834534 | 123 | |
| BC059835.1 | - | 19 - 20 | 2203 | 6406755 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ulk1
Genomic Map