Assay Details
| Gene Symbol: Nr1h2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285517.1 | NP_001272446.1 | 7 - 8 | 1191 | 103 | |
| NM_001285518.1 | NP_001272447.1 | 7 - 8 | 1185 | 103 | ||
| NM_001285519.1 | NP_001272448.1 | 7 - 8 | 1182 | 103 | ||
| NM_009473.3 | NP_033499.1 | 7 - 8 | 1198 | 103 | ||
| GenBank mRNA | AK077620.1 | - | 7 - 8 | 1164 | 103 | |
| AK134489.1 | - | 7 - 8 | 1148 | 103 | ||
| AK156777.1 | - | 7 - 8 | 1166 | 103 | ||
| BC066025.1 | - | 7 - 8 | 1149 | 6829527 | 103 | |
| BC106148.1 | - | 6 - 7 | 918 | 103 | ||
| EU869275.2 | - | 4 - 5 | 1177 | 103 | ||
| U09419.1 | - | 6 - 7 | 1016 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nr1h2
Genomic Map