Assay Details
| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009504.4 | NP_033530.2 | 6 - 7 | 707 | 86 | |
| GenBank mRNA | AK030339.1 | - | 6 - 7 | 708 | 86 | |
| AK154647.1 | - | 6 - 7 | 682 | 86 | ||
| AK156789.1 | - | 6 - 7 | 723 | 86 | ||
| BC006716.1 | - | 6 - 7 | 673 | 3710866 | 86 | |
| CT010294.1 | - | 4 - 5 | 567 | 86 | ||
| D31969.1 | - | 6 - 7 | 675 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vdr
Genomic Map