Assay Details
| Gene Symbol: Vegfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025250.3 | NP_001020421.2 | 1 - 2 | 1087 | 77 | |
| NM_001025257.3 | NP_001020428.2 | 1 - 2 | 1087 | 77 | ||
| NM_001287056.1 | NP_001273985.1 | 1 - 2 | 1087 | 77 | ||
| NM_001287057.1 | NP_001273986.1 | 1 - 2 | 1087 | 77 | ||
| NM_001287058.1 | NP_001273987.1 | 1 - 2 | 1087 | 77 | ||
| NM_009505.4 | NP_033531.3 | 1 - 2 | 1087 | 77 | ||
| GenBank mRNA | AY263146.1 | - | 1 - 2 | 66 | 77 | |
| AY707864.1 | - | 2 - 3 | 284 | 77 | ||
| AY750956.1 | - | 2 - 3 | 245 | 77 | ||
| AY750957.1 | - | 2 - 3 | 208 | 77 | ||
| BC061468.1 | - | 1 - 2 | 460 | 6816435 | 77 | |
| CA321456.1 | - | 1 - 2 | 460 | 77 | ||
| EF028705.1 | - | 2 - 3 | 280 | 77 | ||
| M95200.1 | - | 1 - 2 | 152 | 77 | ||
| S37052.1 | - | 1 - 2 | 78 | 77 | ||
| S38083.1 | - | 1 - 2 | 78 | 77 | ||
| S38100.1 | - | 1 - 2 | 78 | 77 | ||
| U50279.1 | - | 1 - 2 | 66 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (27)
Parathyroid hormone-related protein promotes epithelial-mesenchymal transition.
- Authors
- Ardura JA, Rayego-Mateos S, Ramila D, Ruiz-Ortega M, Esbrit P
- Journal
- J Am Soc Nephrol - Page No 237 (Mon Feb 01 00:00:00 EST 2010)
- Volume (Year)
- 21 (2010)
- Product Usage
- -
Mechanisms of sex differences in TNFR2-mediated cardioprotection.
- Authors
- Wang M, Crisostomo PR, Markel TA, Wang Y, Meldrum DR
- Journal
- Circulation - Page No S38 (Tue Sep 30 00:00:00 EDT 2008)
- Volume (Year)
- 118 (2008)
- Product Usage
- -
Oral iron chelator deferiprone protects against iron overload induced retinal degeneration.
- Authors
- Hadziahmetovic M, Song Y, Wolkow N, Iacovelli J, Grieco S, Lee J, Lyubarsky A, Pratico D, Connelly J, Spino M, Harris ZL, Dunaief J
- Journal
- Invest Ophthalmol Vis Sci - Page No (Thu Nov 04 00:00:00 EDT 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
Reduction in IkappaB kinase alpha expression promotes the development of skin papillomas and carcinomas.
- Authors
- Park E, Zhu F, Liu B, Xia X, Shen J, Bustos T, Fischer SM, Hu Y
- Journal
- Cancer Res - Page No 9158 (Mon Oct 01 00:00:00 EDT 2007)
- Volume (Year)
- 67 (2007)
- Product Usage
- -
Specific blockade of VEGF and HER2 pathways results in greater growth inhibition of breast cancer xenografts that overexpress HER2.
- Authors
- Le XF, Mao W, Lu C, Thornton A, Heymach JV, Sood AK, Bast RC Jr
- Journal
- Cell Cycle - Page No 3747 (Mon Dec 01 00:00:00 EST 2008)
- Volume (Year)
- 7 (2008)
- Product Usage
- -
Early organ-specific endothelial activation during hemorrhagic shock and resuscitation.
- Authors
- van Meurs M, Wulfert FM, Knol AJ, De Haes A, Houwertjes M, Aarts LP, Molema G
- Journal
- Shock - Page No 291 (Fri Feb 01 00:00:00 EST 2008)
- Volume (Year)
- 29 (2008)
- Product Usage
- -
Soleus, plantaris and gastrocnemius VEGF mRNA responses to hypoxia and exercise are preserved in aged compared with young female C57BL/6 mice.
- Authors
- Gavin TP, Westerkamp LM, Zwetsloot KA
- Journal
- Acta Physiol (Oxf) - Page No 113 (Sun Oct 01 00:00:00 EDT 2006)
- Volume (Year)
- 188 (2006)
- Product Usage
- -
VEGFR-3/Flt-4 mediates proliferation and chemotaxis in glial precursor cells.
- Authors
- Kranich S, Hattermann K, Specht A, Lucius R, Mentlein R
- Journal
- Neurochem Int - Page No 747 (Tue Dec 01 00:00:00 EST 2009)
- Volume (Year)
- 55 (2009)
- Product Usage
- -
Leukemia inhibitory factor regulates microvessel density by modulating oxygen-dependent VEGF expression in mice.
- Authors
- Kubota Y, Hirashima M, Kishi K, Stewart CL, Suda T
- Journal
- J Clin Invest - Page No 2393 (Tue Jul 01 00:00:00 EDT 2008)
- Volume (Year)
- 118 (2008)
- Product Usage
- -
Fibroblast growth factor expression during skeletal fracture healing in mice.
- Authors
- Schmid GJ, Kobayashi C, Sandell LJ, Ornitz DM
- Journal
- Dev Dyn - Page No 766 (Sun Mar 01 00:00:00 EST 2009)
- Volume (Year)
- 238 (2009)
- Product Usage
- -
Parathyroid hormone-related protein interacts with vascular endothelial growth factor to promote fibrogenesis in the obstructed mouse kidney.
- Authors
- Ardura JA, Berruguete R, Ramila D, Alvarez-Arroyo MV, Esbrit P
- Journal
- Am J Physiol Renal Physiol - Page No F415 (Fri Aug 01 00:00:00 EDT 2008)
- Volume (Year)
- 295 (2008)
- Product Usage
- -
Topical delivery of silver nanoparticles promotes wound healing.
- Authors
- Tian J, Wong KK, Ho CM, Lok CN, Yu WY, Che CM, Chiu JF, Tam PK
- Journal
- Chemmedchem - Page No 129 (Mon Jan 01 00:00:00 EST 2007)
- Volume (Year)
- 2 (2007)
- Product Usage
- -
Regulation of angiopoietin expression by bacterial lipopolysaccharide.
- Authors
- Mofarrahi M, Nouh T, Qureshi S, Guillot L, Mayaki D, Hussain SN
- Journal
- Am J Physiol Lung Cell Mol Physiol - Page No L955 (Thu May 01 00:00:00 EDT 2008)
- Volume (Year)
- 294 (2008)
- Product Usage
- -
Hypoxia up-regulates expression of Eph receptors and ephrins in mouse skin.
- Authors
- Vihanto MM, Plock J, Erni D, Frey BM, Frey FJ, Huynh-Do U
- Journal
- Faseb J - Page No 1689 (Sat Oct 01 00:00:00 EDT 2005)
- Volume (Year)
- 19 (2005)
- Product Usage
- -
Anti-tumor effects of anti-VEGF siRNA encapsulated with PLGA microspheres in mice.
- Authors
- Murata N, Takashima Y, Toyoshima K, Yamamoto M, Okada H
- Journal
- J Control Release - Page No 246 (Thu Mar 20 00:00:00 EDT 2008)
- Volume (Year)
- 126 (2008)
- Product Usage
- -
von Hippel-Lindau protein regulates transition from the fetal to the adult circulatory system in retina.
- Authors
- Kurihara T, Kubota Y, Ozawa Y, Takubo K, Noda K, Simon MC, Johnson RS, Suematsu M, Tsubota K, Ishida S, Goda N, Suda T, Okano H
- Journal
- Development - Page No 1563 (Sat May 01 00:00:00 EDT 2010)
- Volume (Year)
- 137 (2010)
- Product Usage
- -
Role of hypoxia in obesity-induced disorders of glucose and lipid metabolism in adipose tissue.
- Authors
- Yin J, Gao Z, He Q, Zhou D, Guo Z, Ye J
- Journal
- Am J Physiol Endocrinol Metab - Page No E333 (Sun Feb 01 00:00:00 EST 2009)
- Volume (Year)
- 296 (2009)
- Product Usage
- -
IFATS collection: Fibroblast growth factor-2-induced hepatocyte growth factor secretion by adipose-derived stromal cells inhibits postinjury fibrogenesis through a c-Jun N-terminal kinase-dependent mechanism.
- Authors
- Suga H, Eto H, Shigeura T, Inoue K, Aoi N, Kato H, Nishimura S, Manabe I, Gonda K, Yoshimura K
- Journal
- Stem Cells - Page No 238 (Thu Jan 01 00:00:00 EST 2009)
- Volume (Year)
- 27 (2009)
- Product Usage
- -
CD147 impacts angiogenesis and metastasis formation.
- Authors
- Voigt H, Vetter-Kauczok CS, Schrama D, Hofmann UB, Becker JC, Houben R
- Journal
- Cancer Invest - Page No 329 (Sun Mar 01 00:00:00 EST 2009)
- Volume (Year)
- 27 (2009)
- Product Usage
- -
Macrophage infiltration into adipose tissue may promote angiogenesis for adipose tissue remodeling in obesity.
- Authors
- Pang C, Gao Z, Yin J, Zhang J, Jia W, Ye J
- Journal
- Am J Physiol Endocrinol Metab - Page No E313 (Fri Aug 01 00:00:00 EDT 2008)
- Volume (Year)
- 295 (2008)
- Product Usage
- -
Critical role for osteopontin in diabetic nephropathy.
- Authors
- Nicholas SB, Liu J, Kim J, Ren Y, Collins AR, Nguyen L, Hsueh WA
- Journal
- Kidney Int - Page No 588 (Thu Apr 01 00:00:00 EDT 2010)
- Volume (Year)
- 77 (2010)
- Product Usage
- -
Functions of paracrine PDGF signaling in the proangiogenic tumor stroma revealed by pharmacological targeting.
- Authors
- Pietras K, Pahler J, Bergers G, Hanahan D
- Journal
- Plos Med - Page No e19 (Tue Jan 29 00:00:00 EST 2008)
- Volume (Year)
- 5 (2008)
- Product Usage
- -
Simvastatin enhances VEGF production and ameliorates impaired wound healing in experimental diabetes.
- Authors
- Bitto A, Minutoli L, Altavilla D, Polito F, Fiumara T, Marini H, Galeano M, Calo M, Lo Cascio P, Bonaiuto M, Migliorato A, Caputi AP, Squadrito F
- Journal
- Pharmacol Res - Page No 159 (Fri Feb 01 00:00:00 EST 2008)
- Volume (Year)
- 57 (2008)
- Product Usage
- -
Efficient regulation of VEGF expression by promoter-targeted lentiviral shRNAs based on epigenetic mechanism: a novel example of epigenetherapy.
- Authors
- Turunen MP, Lehtola T, Heinonen SE, Assefa GS, Korpisalo P, Girnary R, Glass CK, Vaisanen S, Yla-Herttuala S
- Journal
- Circ Res - Page No 604 (Fri Sep 11 00:00:00 EDT 2009)
- Volume (Year)
- 105 (2009)
- Product Usage
- -
Histone deacetylase inhibition reduces myocardial ischemia-reperfusion injury in mice.
- Authors
- Granger A, Abdullah I, Huebner F, Stout A, Wang T, Huebner T, Epstein JA, Gruber PJ
- Journal
- Faseb J - Page No 3549 (Wed Oct 01 00:00:00 EDT 2008)
- Volume (Year)
- 22 (2008)
- Product Usage
- -
Deletion of Forkhead Box M1 transcription factor from respiratory epithelial cells inhibits pulmonary tumorigenesis.
- Authors
- Wang IC, Meliton L, Ren X, Zhang Y, Balli D, Snyder J, Whitsett JA, Kalinichenko VV, Kalin TV
- Journal
- Plos One - Page No e6609 (Thu Jan 01 00:00:00 EST 2009)
- Volume (Year)
- 4 (2009)
- Product Usage
- -
Sema3a maintains normal heart rhythm through sympathetic innervation patterning.
- Authors
- Ieda M, Kanazawa H, Kimura K, Hattori F, Ieda Y, Taniguchi M, Lee JK, Matsumura K, Tomita Y, Miyoshi S, Shimoda K, Makino S, Sano M, Kodama I, Ogawa S, Fukuda K
- Journal
- Nat Med - Page No 604 (Tue May 01 00:00:00 EDT 2007)
- Volume (Year)
- 13 (2007)
- Product Usage
- -

Vegfa
Genomic Map