Assay Details
Gene Symbol: Wnt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009518.2 | NP_033544.1 | 1 - 2 | 268 | 72 | |
GenBank mRNA | BC014737.1 | - | 1 - 2 | 268 | 4921327 | 72 |
U61969.1 | - | 778 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
