Assay Details
| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290350.1 | NP_001277279.1 | 4 | 871 | 64 | |
| NM_001290351.1 | NP_001277280.1 | 4 | 862 | 64 | ||
| NM_001290353.1 | NP_001277282.1 | 4 | 769 | 64 | ||
| NM_009554.4 | NP_033580.3 | 4 | 859 | 64 | ||
| GenBank mRNA | AK032469.1 | - | 4 | 741 | 64 | |
| BC024863.1 | - | 2 | 482 | 64 | ||
| BC063757.1 | - | 4 | 743 | 6331110 | 64 | |
| X52533.1 | - | 1 | 157 | 64 | ||
| X64413.1 | - | 4 | 665 | 64 | ||
| X89264.1 | - | 5 | 771 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963159 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963157 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 511023 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 677002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409341 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | AK041747.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | AK045343.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | AK080671.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | AK084226.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | AK147045.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | BC049253.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | BC094315.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 781088 | NM_178724.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65818 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112284 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P165215 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P190454 | X89264.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P190454 | X64413.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P190454 | BC063757.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P190454 | X52533.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P190454 | BC024863.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22963159 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112284 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P165215 | - | - |
| Whole Mouse Genome G4122A | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P223269 | AK032469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190454 | X89264.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190454 | X64413.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190454 | BC063757.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190454 | X52533.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P190454 | BC024863.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123192 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P165215 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03685085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03685090 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03685088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03685087 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963157 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22963159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22963082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03685094 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03976675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03282170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963163 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03495086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963161 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963122 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03856223 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963167 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03263958 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963125 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963173 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963164 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03685076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963121 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03543141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03685086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03311287 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03799792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22963175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10513550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513546 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513545 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10513549 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449391_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419207_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10513544 | - | - |
| Mouse Genome 430 2.0 Array | 1419207_at | - | - |
| Mouse Genome 430 2.0 Array | 1449391_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419207_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449391_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22696_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 22696_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400022768.mm.1 | TC0400002782.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400022768.mm.1 | NM_009554.4 | Refseq |
| Mouse Expression Set 430 | 1449391_PM_at | - | - |
| Mouse Expression Set 430 | 1449391_at | - | - |
| Mouse Expression Set 430 | 1419207_at | - | - |
| Mouse Expression Set 430 | 1419207_PM_at | - | - |
| Murine Genome U74v2 Set | 161605_PM_f_at | - | - |
| Murine Genome U74v2 Set | 161605_f_at | - | - |
| Murine Genome U74v2 Set | 92974_PM_at | - | - |
| Murine Genome U74v2 Set | 92974_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449391_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419207_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419207_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449391_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419207_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419207_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449391_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449391_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 574875 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102340347 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 7000722 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102340347 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102030309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0007000722 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2479897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2479897 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2576801 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1220542 | - | - |
| mouse-6 v1.1 (Illumina) | scl24268.3_61-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0022696.2_206-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 7000722 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102030309 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102340347 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0022696.2_206-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102340347 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102030309 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7000722 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2809334 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2479899 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2809334 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2576801 | - | - |

Zfp37
Genomic Map