Assay Details
| Gene Symbol: Map3k12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163643.1 | NP_001157115.1 | 9 - 10 | 1673 | 96 | |
| NM_009582.4 | NP_033608.3 | 7 - 8 | 1497 | 96 | ||
| GenBank mRNA | AK029882.1 | - | 7 - 8 | 1306 | 96 | |
| AK034374.1 | - | 9 - 10 | 1489 | 96 | ||
| AK081623.1 | - | 2 - 3 | 165 | 96 | ||
| AK166435.1 | - | 9 - 10 | 1593 | 96 | ||
| BC047158.1 | - | 6 - 7 | 1291 | 6494216 | 96 | |
| BC057572.1 | - | 9 - 10 | 1497 | 6401321 | 96 | |
| U14636.1 | - | 7 - 8 | 1329 | 96 | ||
| U23789.1 | - | 6 - 7 | 1287 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map3k12
Genomic Map