Assay Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009612.3 | NP_033742.2 | 2 - 3 | 201 | 89 | |
| GenBank mRNA | L48015.1 | - | 2 - 3 | 203 | 89 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the L48015.1,NM_009612.3 transcripts.
Set Membership: |
|

Acvrl1
Genomic Map