Assay Details
| Gene Symbol: Acvrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277255.1 | NP_001264184.1 | 9 - 10 | 1336 | 77 | |
| NM_001277257.1 | NP_001264186.1 | 8 - 9 | 1275 | 77 | ||
| NM_001277258.1 | NP_001264187.1 | 8 - 9 | 1244 | 77 | ||
| NM_001277259.1 | NP_001264188.1 | 9 - 10 | 1331 | 77 | ||
| NM_009612.3 | NP_033742.2 | 8 - 9 | 1313 | 77 | ||
| GenBank mRNA | AK134712.1 | - | 6 - 7 | 1070 | 77 | |
| AK160915.1 | - | 9 - 10 | 1304 | 77 | ||
| AK170870.1 | - | 6 - 7 | 1069 | 77 | ||
| BC014291.1 | - | 7 - 8 | 1232 | 3982004 | 77 | |
| BC015083.1 | - | 7 - 8 | 1194 | 3980151 | 77 | |
| CT010330.1 | - | 6 - 7 | 1041 | 77 | ||
| L48015.1 | - | 8 - 9 | 1315 | 77 | ||
More Information
Set Membership: |
|

Acvrl1
Genomic Map