Assay Details
| Gene Symbol: Aqp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009699.3 | NP_033829.3 | 2 - 3 | 616 | 69 | |
| GenBank mRNA | AF020519.1 | - | 2 - 3 | 519 | 69 | |
| AK002689.1 | - | 2 - 3 | 69 | |||
| AK142567.1 | - | 2 - 3 | 617 | 69 | ||
| BC019966.1 | - | 2 - 3 | 600 | 4222942 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aqp2
Genomic Map