Assay Details
| Gene Symbol: Areg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009704.3 | NP_033834.1 | 3 - 4 | 696 | 76 | |
| GenBank mRNA | AK018590.1 | - | 3 - 4 | 712 | 76 | |
| AK020273.1 | - | 3 - 4 | 712 | 76 | ||
| BC009138.1 | - | 3 - 4 | 696 | 3597695 | 76 | |
| CT010320.1 | - | 3 - 4 | 497 | 76 | ||
| D12648.1 | - | 3 - 4 | 523 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Areg
Genomic Map