Assay Details
| Gene Symbol: Atp2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036684.2 | NP_001031761.1 | 9 - 10 | 1958 | 58 | |
| NM_009723.5 | NP_033853.1 | 11 - 12 | 2147 | 58 | ||
| GenBank mRNA | AB030737.1 | - | 8 - 9 | 1519 | 58 | |
| AB030738.1 | - | 8 - 9 | 1519 | 58 | ||
| AF053471.1 | - | 10 - 11 | 1976 | 58 | ||
| AK054112.1 | - | 13 - 14 | 2245 | 58 | ||
| AK147240.1 | - | 11 - 12 | 2033 | 58 | ||
| AK147357.1 | - | 9 - 10 | 1887 | 58 | ||
| AK147369.1 | - | 11 - 12 | 2083 | 58 | ||
| AK147399.1 | - | 9 - 10 | 2122 | 58 | ||
| AK147501.1 | - | 11 - 12 | 2094 | 58 | ||
| BC075643.1 | - | 9 - 10 | 1958 | 30621322 | 58 | |
| GU734816.2 | - | 11 - 12 | 1654 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2b2
Genomic Map