Assay Details
| Gene Symbol: Atp2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036684.2 | NP_001031761.1 | 10 - 11 | 2209 | 56 | |
| NM_009723.5 | NP_033853.1 | 12 - 13 | 2398 | 56 | ||
| GenBank mRNA | AB030737.1 | - | 9 - 10 | 1770 | 56 | |
| AB030738.1 | - | 9 - 10 | 1770 | 56 | ||
| AF053471.1 | - | 11 - 12 | 2227 | 56 | ||
| AK054112.1 | - | 14 - 15 | 2496 | 56 | ||
| AK147240.1 | - | 12 - 13 | 2284 | 56 | ||
| AK147357.1 | - | 10 - 11 | 2138 | 56 | ||
| AK147369.1 | - | 12 - 13 | 2333 | 56 | ||
| AK147399.1 | - | 10 - 11 | 2373 | 56 | ||
| AK147501.1 | - | 12 - 13 | 2345 | 56 | ||
| GU734816.2 | - | 12 - 13 | 1905 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2b2
Genomic Map