Assay Details
| Gene Symbol: C4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009780.2 | NP_033910.2 | 17 - 18 | 2330 | 73 | |
| GenBank mRNA | AK128974.1 | - | 5 - 6 | 756 | 73 | |
| AK158256.1 | - | 17 - 18 | 2316 | 73 | ||
| AK172637.1 | - | 17 - 18 | 2319 | 73 | ||
| BC039141.1 | - | 17 - 18 | 2293 | 73 | ||
| BC055297.1 | - | 17 - 18 | 2300 | 73 | ||
| BC067394.1 | - | 17 - 18 | 2300 | 30107263 | 73 | |
| BC067409.1 | - | 17 - 18 | 2300 | 30098613 | 73 | |
| BC150743.1 | - | 17 - 18 | 2280 | 9087654 | 73 | |
| BC158005.1 | - | 17 - 18 | 2279 | 9007313 | 73 | |
| M11729.1 | - | 17 - 18 | 2315 | 73 | ||
| M12970.1 | - | 309 | 73 | |||
| X05314.1 | - | 4 - 5 | 489 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C4b
Genomic Map