Assay Details
| Gene Symbol: Camk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009793.3 | NP_033923.2 | 5 - 6 | 796 | 69 | |
| GenBank mRNA | AK030219.1 | - | 5 - 6 | 680 | 69 | |
| AK043114.1 | - | 5 - 6 | 506 | 69 | ||
| BC070420.1 | - | 5 - 6 | 677 | 5718423 | 69 | |
| X58995.1 | - | 5 - 6 | 486 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Camk4
Genomic Map