Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009808.4 | NP_033938.3 | 5 - 6 | 530 | 80 | |
| GenBank mRNA | AK030309.1 | - | 5 - 6 | 505 | 80 | |
| AK046681.1 | - | 5 - 6 | 529 | 80 | ||
| AK053407.1 | - | 5 - 6 | 510 | 80 | ||
| AK077256.1 | - | 5 - 6 | 532 | 80 | ||
| AK080045.1 | - | 5 - 6 | 531 | 80 | ||
| AK161525.1 | - | 5 - 6 | 516 | 80 | ||
| AY675223.1 | - | 5 - 6 | 292 | 80 | ||
| AY675224.1 | - | 5 - 6 | 504 | 80 | ||
| BC028979.1 | - | 5 - 6 | 517 | 5375109 | 80 | |
| Y13090.1 | - | 4 - 5 | 520 | 80 | ||
More Information
Set Membership: |
|

Casp12
Genomic Map