Assay Details
| Gene Symbol: Casp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009808.4 | NP_033938.3 | 7 - 8 | 941 | 87 | |
| GenBank mRNA | AK030309.1 | - | 7 - 8 | 916 | 87 | |
| AK077256.1 | - | 7 - 8 | 943 | 87 | ||
| AK080045.1 | - | 7 - 8 | 953 | 87 | ||
| AK161525.1 | - | 7 - 8 | 927 | 87 | ||
| AY675223.1 | - | 7 - 8 | 703 | 87 | ||
| AY675224.1 | - | 7 - 8 | 915 | 87 | ||
| BC028979.1 | - | 7 - 8 | 928 | 5375109 | 87 | |
| Y13090.1 | - | 6 - 7 | 931 | 87 | ||
More Information
Set Membership: |
|

Casp12
Genomic Map