Assay Details
| Gene Symbol: Casp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009809.5 | NP_033939.1 | 3 - 4 | 404 | 88 | |
| GenBank mRNA | AF092997.1 | - | 2 - 3 | 249 | 88 | |
| AJ007750.1 | - | 3 - 4 | 269 | 88 | ||
| AK081645.1 | - | 3 - 4 | 357 | 88 | ||
| BC101959.1 | - | 3 - 4 | 342 | 40044873 | 88 | |
| BC101960.1 | - | 3 - 4 | 342 | 40044874 | 88 | |
| BC103555.1 | - | 3 - 4 | 342 | 40044870 | 88 | |
| BC103556.1 | - | 3 - 4 | 342 | 40044871 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casp14
Genomic Map