Assay Details
| Gene Symbol: Cckar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009827.2 | NP_033957.1 | 1 - 2 | 465 | 92 | |
| GenBank mRNA | AK004730.2 | - | 1 - 2 | 323 | 92 | |
| AK143895.1 | - | 1 - 2 | 313 | 92 | ||
| AK146738.1 | - | 1 - 2 | 466 | 92 | ||
| AK147052.1 | - | 1 - 2 | 466 | 92 | ||
| BC020534.1 | - | 1 - 2 | 285 | 4236240 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cckar
Genomic Map