Assay Details
| Gene Symbol: Cckar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009827.2 | NP_033957.1 | 3 - 4 | 972 | 64 | |
| GenBank mRNA | AK004730.2 | - | 3 - 4 | 830 | 64 | |
| AK143895.1 | - | 3 - 4 | 820 | 64 | ||
| AK146738.1 | - | 3 - 4 | 972 | 64 | ||
| AK147052.1 | - | 3 - 4 | 972 | 64 | ||
| AK164301.1 | - | 564 | 64 | |||
| BC020534.1 | - | 3 - 4 | 792 | 4236240 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cckar
Genomic Map