Assay Details
| Gene Symbol: Ccna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009828.2 | NP_033958.2 | 2 - 3 | 605 | 90 | |
| GenBank mRNA | AK044924.1 | - | 2 - 3 | 608 | 90 | |
| AK135861.1 | - | 2 - 3 | 577 | 90 | ||
| AK216847.1 | - | 2 - 3 | 277 | 90 | ||
| BC052730.1 | - | 2 - 3 | 553 | 6816226 | 90 | |
| X75483.1 | - | 2 - 3 | 553 | 90 | ||
| Z26580.1 | - | 2 - 3 | 562 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cyclin-dependent protein serine/threonine kinase activity
cell cycle
mitotic nuclear division
regulation of G2/M transition of mitotic cell cycle
positive regulation of transcription, DNA-templated
positive regulation of fibroblast proliferation
cell division
protein binding
protein kinase binding
regulation of cyclin-dependent protein serine/threonine kinase activity
cell cycle
mitotic nuclear division
regulation of G2/M transition of mitotic cell cycle
positive regulation of transcription, DNA-templated
positive regulation of fibroblast proliferation
cell division
protein binding
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058854 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058861 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270083 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411520 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880539 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150923 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058859 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058856 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22058848 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04874255 | - | - |
| Whole Mouse Genome G4122A | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | Z26580.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | BC052730.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | NM_009828.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P481920 | AK044924.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128819 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134944 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02711956 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02711950 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03098243 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02711955 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058859 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058853 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058861 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058856 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058860 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058857 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058858 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058855 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058863 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058862 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22058854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058825 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22058835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058812 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058838 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058837 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058814 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02711950 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02711948 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058803 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058819 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058830 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058865 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058797 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02711955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02711952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058823 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058840 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058816 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058807 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058829 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02711956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058835 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22058864 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10497833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491563 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10491562 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10497835 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417910_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417911_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10497831 | - | - |
| Mouse Genome 430 2.0 Array | 1417911_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417911_at | - | - |
| Mouse Genome 430 2.0 Array | 1417910_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417910_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12428_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12428_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300014363.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014363.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014363.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014355.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014355.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014355.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014357.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014357.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014357.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014360.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014360.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014360.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014353.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014353.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014353.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014354.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014354.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014354.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014364.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014364.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014364.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014367.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014367.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014367.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014352.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014352.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014352.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014359.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014359.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014359.mm.1 | TC0300001856.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300014358.mm.1 | NM_009828.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014358.mm.1 | NM_009828.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300014358.mm.1 | TC0300001856.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417910_at | - | - |
| Mouse Expression Set 430 | 1417911_at | - | - |
| Mouse Expression Set 430 | 1417911_PM_at | - | - |
| Mouse Expression Set 430 | 1417910_PM_at | - | - |
| Murine Genome U74v2 Set | 99186_PM_at | - | - |
| Murine Genome U74v2 Set | 99186_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417911_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417911_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417910_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417910_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417910_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417910_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417911_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417911_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5290075 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5290075 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005290075 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2672442 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2672442 | - | - |
| mouse-6 v1.1 (Illumina) | scl22236.8_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5290075 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl22236.8_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5290075 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2672442 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2672442 | - | - |
Citations (1)
Hyaluronan and CD44 antagonize mitogen-dependent cyclin D1 expression in mesenchymal cells.
- Authors
- Kothapalli D, Zhao L, Hawthorne EA, Cheng Y, Lee E, Pure E, Assoian RK
- Journal
- J Cell Biol - Page No 535 (Mon Feb 12 00:00:00 EST 2007)
- Volume (Year)
- 176 (2007)
- Product Usage
- -

Ccna2
Genomic Map