Assay Details
| Gene Symbol: Ccng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009831.2 | NP_033961.1 | 3 - 4 | 796 | 104 | |
| GenBank mRNA | AK128957.1 | - | 3 - 4 | 784 | 104 | |
| AK136645.1 | - | 3 - 4 | 533 | 104 | ||
| AK143114.1 | - | 3 - 4 | 787 | 104 | ||
| AK147026.1 | - | 3 - 4 | 784 | 104 | ||
| AK153392.1 | - | 3 - 4 | 728 | 104 | ||
| AK159429.1 | - | 3 - 4 | 726 | 104 | ||
| AK159831.1 | - | 3 - 4 | 731 | 104 | ||
| AK160411.1 | - | 3 - 4 | 771 | 104 | ||
| AK167052.1 | - | 3 - 4 | 728 | 104 | ||
| AK167486.1 | - | 3 - 4 | 723 | 104 | ||
| AK170397.1 | - | 3 - 4 | 788 | 104 | ||
| AK178990.1 | - | 2 - 3 | 309 | 104 | ||
| AK209730.1 | - | 1 - 2 | 250 | 104 | ||
| BC005534.1 | - | 2 - 3 | 446 | 104 | ||
| L49507.1 | - | 3 - 4 | 739 | 104 | ||
| Z37110.1 | - | 3 - 4 | 739 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccng1
Genomic Map