Assay Details
| Gene Symbol: Cd8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009858.2 | NP_033988.1 | 2 - 3 | 456 | 77 | |
| GenBank mRNA | AK087944.1 | - | 2 - 3 | 433 | 77 | |
| AK088372.1 | - | 2 - 3 | 481 | 77 | ||
| AK088649.1 | - | 2 - 3 | 480 | 77 | ||
| AK154055.1 | - | 2 - 3 | 436 | 77 | ||
| AK162993.1 | - | 2 - 3 | 445 | 77 | ||
| AK169581.1 | - | 2 - 3 | 457 | 77 | ||
| M16799.1 | - | 2 - 3 | 449 | 77 | ||
| M17534.1 | - | 2 - 3 | 456 | 77 | ||
| M19504.1 | - | 2 - 3 | 444 | 77 | ||
| X07698.1 | - | 2 - 3 | 460 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd8b1
Genomic Map