Assay Details
| Gene Symbol: Cdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271567.1 | NP_001258496.1 | 1 - 2 | 104 | 85 | |
| NM_001271568.1 | NP_001258497.1 | 1 - 2 | 104 | 85 | ||
| NM_009863.3 | NP_033993.2 | 1 - 2 | 104 | 85 | ||
| GenBank mRNA | AB018574.1 | - | 1 - 2 | 104 | 86 | |
| AB018575.1 | - | 1 - 2 | 104 | 86 | ||
| AJ007661.1 | - | 1 - 2 | 56 | 85 | ||
| AK012986.1 | - | 1 - 2 | 87 | 85 | ||
| AK145921.1 | - | 1 - 2 | 30 | 85 | ||
| AK163921.1 | - | 74 | 85 | |||
| BC062149.1 | - | 1 - 2 | 34 | 85 | ||
| BC072666.1 | - | 1 - 2 | 45 | 30611704 | 85 | |
| BC080702.1 | - | 1 - 2 | 57 | 30474846 | 85 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB018574.1,AB018575.1,AJ007661.1,AK012986.1,AK145921.1,AK163921.1,BC072666.1,BC080702.1,NM_001271567.1,NM_001271568.1,NM_009863.3 transcripts.
Set Membership: |
|

Cdc7
Genomic Map