Assay Details
| Gene Symbol: Cdkn1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161624.1 | NP_001155096.1 | 2 - 3 | 1118 | 115 | |
| NM_009876.4 | NP_034006.3 | 2 - 3 | 1080 | 115 | ||
| GenBank mRNA | AK077844.1 | - | 2 - 3 | 1029 | 115 | |
| AK132278.1 | - | 2 - 3 | 1027 | 115 | ||
| BC005412.1 | - | 2 - 3 | 1017 | 3490380 | 115 | |
| U20553.1 | - | 2 - 3 | 951 | 115 | ||
| U22399.1 | - | 2 - 3 | 1009 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdkn1c
Genomic Map